missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMMR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15820-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Antibodies raised against RHAMM have been reported to recognize proteins of 56, 66 and 70 kDa in the supernatant of murine 3T3 fibroblasts. Some of the isoforms detectable by anti-RHAMM antibodies, including the 52 kDa form and a 48-49 kDa form found in 1
The protein encoded by this gene is involved in cell motility. It is expressed in breast tissue and together with other proteins, it forms a complex with BRCA1 and BRCA2, thus is potentially associated with higher risk of breast cancer. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.Especificaciones
| HMMR | |
| Polyclonal | |
| Unconjugated | |
| HMMR | |
| CD168, HMMR, IHABP, RHAMM | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15366, 25460, 3161 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75330, P97779, Q00547 | |
| HMMR | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido