missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HMGB1/HMGB2 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 16326714
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16326714 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16326714 Proveedor Invitrogen™ N.º de proveedor PA578005

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2, A375, mouse brain, rat brain Predicted Reactivity: Xenopus laevis (87%), Pig (99%), Chicken (92%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

HMGB1/HMGB2 encodes a member of the non-histone chromosomal high mobility group protein family. The proteins of this family are chromatin-associated and ubiquitously distributed in the nucleus of higher eukaryotic cells. In vitro studies have demonstrated that this protein is able to efficiently bend DNA and form DNA circles. These studies suggest a role in facilitating cooperative interactions between cis-acting proteins by promoting DNA flexibility. This protein was also reported to be involved in the final ligation step in DNA end-joining processes of DNA double-strand breaks repair and V(D)J recombination.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno HMGB1/HMGB2
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 20% glycerol and 0.01% thimerosal; pH 7
génica HMGB1
N.º de referencia del gen P09429, P26583, P30681, P52925, P63158, P63159
Alias de gen Ac2-008; amphoterin; C80539; DEF; heparin-binding protein p30; high mobility group 1; high mobility group box 1; high mobility group box 2; high mobility group protein 1; High mobility group protein 2; high mobility group protein B1; High mobility group protein B2; high-mobility group (nonhistone chromosomal) protein 1; high-mobility group (nonhistone chromosomal) protein 2; high-mobility group box 1; high-mobility group box 2; HMG1; HMG-1; HMG2; HMG-2; HMG3; Hmgb1; Hmgb2; p30; SBP-1; Sulfoglucuronyl carbohydrate binding protein
Símbolos de los genes HMGB1, HMGB2
Especie del huésped Rabbit
Inmunógeno Full length human HMGB1 + HMGB2 Recombinant protein.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 15289, 25459, 29395, 3146, 3148, 97165
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.