Descripción
Plays a role in neuronal differentiation as chromatin-associated protein. Acts as inhibitor of HMG20B. Overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. Involved in the recruitment of the histone methyltransferase KMT2A/MLL1 and consequent increased methylation of histone H3 lysine 4.
Especificaciones
Especificaciones
| Antígeno | HMG20A |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.25 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | HMG20A |
| N.º de referencia del gen | Q9DC33, Q9NP66 |
| Alias de gen | high mobility group 20A, HMG box containing protein 20A, HMG20A, HMGX1, HMGXB1 |
| Símbolos de los genes | Hmg20a |
| Mostrar más |