missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLCS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14046-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene shares strong similarity with Saccharomyces cerevisiae Cdc23, a protein essential for cell cycle progression through the G2/M transition. This protein is a component of anaphase-promoting complex (APC), which is composed of eight protein subunits and highly conserved in eukaryotic cells. APC catalyzes the formation of cyclin B-ubiquitin conjugate that is responsible for the ubiquitin-mediated proteolysis of B-type cyclins. This protein and 3 other members of the APC complex contain the TPR (tetratricopeptide repeat), a protein domain important for protein-protein interaction.Especificaciones
| HLCS | |
| Polyclonal | |
| Unconjugated | |
| HLCS | |
| Biotin protein ligase, HCS, HLCS, Holocarboxylase synthetase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110948, 288240, 3141 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50747, Q920N2 | |
| HLCS | |
| HLCS Fusion Protein Ag5208 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido