missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HLA-DRw52 Monoclonal Antibody (7.3.19.1)
GREENER_CHOICE

Código de producto. 11530852
Change view
Click to view available options
Cantidad:
1 mL
Tamaño de la unidad:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11530852 1 mL 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11530852 Proveedor Invitrogen™ N.º de proveedor MA135422

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The antibody reacts with a polymorphic determinant on HLA-DR molecules which is associated with DR3, DR5, and DRw6 (DRw52-like). It does not react with DRw8. The antibody reacts with part of the HLA-D molecules (29+34 kD). The determinant is strongly expressed on DR3 positive cells and more weakly on DR5 and DRw6 positive cells. The antibody is useful for HLA-DR typing and biochemical characterization of the DR52w positive molecules and functional studies. Cross reacts with Chimpanzee, Macaca-apes and gorilla.

HLA DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of an alpha chain (36 kDa) and a beta chain (27 kDa). It is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides and present them to T-helper cells. They therefore have a critical role in the initiation of the immune response. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno HLA-DRw52
Aplicaciones Cytotoxicity Assay, Immunocytochemistry
Clasificación Monoclonal
Clon 7.3.19.1
Concentración Conc. Not Determined
Conjugado Unconjugated
Formulación PBS with 0.1% BSA and 0.02% sodium azide
génica HLA-DRB1
N.º de referencia del gen P01911
Alias de gen Clone P2-beta-3; DR-16; DRB1; DRw10; DW2.2/DR2.2; FLJ75017; FLJ76359; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA-DR1B; HLA-DRB; HLA-DRB1; HLA-DRB2; human leucocyte antigen DRB1; lymphocyte antigen DRB1; major histocompatibility complex, class II, DR beta 1; MHC class II antigen; MHC class II antigen DRB1*15; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; MHC class II HLA-DR beta 1 chain; MHC class II HLA-DR-beta cell surface glycoprotein; MHC class II HLA-DRw10-beta; SS1; XXbac-BPG161M6.1
Símbolos de los genes HLA-DRB1
Especie del huésped Mouse
Inmunógeno Human HLA-DRw52.
Cantidad 1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3123
Especies diana Human, Non-human Primate
Contenido y almacenamiento 4°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.