Descripción
Expression of the invariant chain of the HLA-DR antigen (CD74) by B-lymphocytes begins in the pre-B stage but lost before the final differentiation to plasma cells. This protein is expressed in B-lymphocytes and many B-cell lymphomas.
Especificaciones
Especificaciones
| Antígeno | HLA-DRA |
| Aplicaciones | Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.21 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | HLA-DRA |
| N.º de referencia del gen | P01903 |
| Alias de gen | HLA DRA, HLA DRA1, HLA-DRA, MHC class II antigen DRA |
| Símbolos de los genes | HLA-DRA |
| Mostrar más |