missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HLA-ABCE Monoclonal Antibody (TP25.99SF), Alexa Fluor™ 488
GREENER_CHOICE

Código de producto. 17604596
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17604596 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17604596 Proveedor Invitrogen™ N.º de proveedor MA544136

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. View return policy

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope on HLA-ABC and HLA-E molecules, but not HLA-G. It can be used for discrimination between HLA-G and other HLA-class I antigens.

HLA-A, -B, and -C are major histocompatibility complex (MHC)-class I antigens. Like other class I molecules (i.e. HLA-E, -F, -G), HLA-A, -B, and -C are heterodimers consisting of a 40 - 45 kDa transmembrane glycoproteic alpha-chain, noncovalently combined to the invariant B2-microglobulin. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno HLA-ABCE
Aplicaciones Flow Cytometry, Western Blot
Clasificación Monoclonal
Clon TP25.99SF
Concentración 0.1 mg/mL
Conjugado Alexa Fluor 488
Formulación PBS with 0.2% BSA and 15mM sodium azide
génica HLA-E
N.º de referencia del gen P01889, P04439, P10321, P13747
Alias de gen PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Símbolos de los genes HLA-A, HLA-B, HLA-C, HLA-E
Especie del huésped Mouse
Inmunógeno IFN-gamma-treated human melanoma cells Colo 38.
Método de purificación Size-exclusion chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3105, 3106, 3107, 3133
Especies diana Human
Contenido y almacenamiento 4°C, store in dark, DO NOT FREEZE!
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.