missing translation for 'onlineSavingsMsg'
Learn More

HLA-A2 Monoclonal Antibody (BB7.2), NovaFluor™ Red 710, eBioscience™, Invitrogen™

Código de producto. p-7250754
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
paquete de 100
paquete de 25
Código de producto. Cantidad unitSize
17843044 25 Tests paquete de 25
17832974 100 Tests paquete de 100
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17843044

Marca: Invitrogen H070T02R04

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

This BB7.2 monoclonal antibody recognizes the human major histocompatibility complex (MHC) class I molecule HLA-A2, which is a member of the HLA-A family. MHC class I exists as a single alpha chain composed of three domains. This molecule associates with beta 2-microglobulin and is expressed on all nucleated cells. The BB7.2 monoclonal antibody has been reported to exhibit blocking activity, as well as recognize HLA-A28. Applications Tested: This BB7.2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5uL (0.6 ug) per test. A test is defined as the amount (ug) of antibody that will stain a cell sample in a final volume of 100uL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. NovaFluor dyes are not compatible with DNA intercalating viability dyes. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes. Use 5uL of CellBlox Blocking Buffer per stained cell sample containing 10^3 to 10^8 cells. Excitation: 639 nm; Emission: 710 nm; Laser: 633-640 nm (Red) Laser

HLA-A belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Hundreds of HLA-A alleles have been described. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno HLA-A2
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon BB7.2
Concentración 0.6 μg/Test
Conjugado NovaFluor Red 710
Formulación PBS with 0.09% sodium azide; pH 7.2
génica HLA-A
N.º de referencia del gen P04439
Alias de gen A29 protein; antigen presenting molecule; Aw-33; Aw-74; DAQB-90C11.16; FLJ26655; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA locus; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; Human leukocyte antigen A; leucocyte antigen A; leukocyte antigen class I-A; major histocompatibility complex, class I, A; MCH class I antigen; MHC 1; MHC class 1 antigen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen null protein; MHC class I antigene; MHC class II antigen; MHC classI antigen; MHC1; truncated MHC class I antigen
Símbolos de los genes HLA-A
Especie del huésped Mouse
Método de purificación Affinity chromatography
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3105
Especies diana Human
Contenido y almacenamiento 4° C, store in dark, DO NOT FREEZE!
Formulario Liquid
Isotype IgG2b κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.