missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIV1 nef Monoclonal Antibody (3E6)

Descripción
This antibody reacts with HIV-1 Nef subtypes A, B and C; BRU, IIIB, LAV and BH10 strains. It has been mapped to amino acids 163-173 (SLLHPMSLHGM - B subtypes consensus). This clone may also be referred to as 3E6 (01-004).
HIV-1 Nef is one of the accessory proteins synthesized in the early stage of AIDS virus reproduction and is abundantly found in infected cells. The name derives from its negative factor thought at the beginning but presently it is remarked as the protein which bears a most distinctive biological characteristic of AIDS virus. The protein interacts directly with the signal transduction protein of the host T cell and works effectively on AIDS infection or on long term survival of the infected cells or induces apoptosis of non-infected cells. It is also involved in the endocytosis and degradation of receptor protein of the cell surface such as CD 4 and MH4, important for AIDS virus infection.
Especificaciones
Especificaciones
| Antígeno | HIV1 nef |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 3E6 |
| Concentración | 1.0 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% sodium azide; pH 7.4 |
| Alias de gen | HIV-1 Nef |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant Nef protein of HIV-1 B subtype consensus. |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?