missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, p Ser28 Antibody (117C826), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB100-56595JF549
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Fish samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones
| Histone H3 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Fish | |
| Purified |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 117C826 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| This Histone H3 [p Ser28] antibody (117C826) was raised against a synthetic peptide containing phospho serine at position 28 CARKS*APATGGVKK of human Histone H3. The immunogen sequence is conserved across mammalian and invertebrates, as well as plants. | |
| 0.1 mL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Store at 4C in the dark. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido