missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIST2H2BE Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA560199
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: KESYSIYVYK VLKQVHPDTG ISSKAM Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Especificaciones
| HIST2H2BE | |
| Polyclonal | |
| Unconjugated | |
| H2BC21 | |
| AV127319; GL105; H2B; H2b 613; H2B histone family, member Q; H2B.1; H2B/q; H2b-613; H2BFQ; H2BGL105; H2BQ; Hist2h2be; HIST2H2BF; histone 2, H2be; histone cluster 2, H2be; histone cluster 2, H2bf; histone H2B type 2-E; Histone H2B type 2-F; Histone H2B.q; histone H2B-GL105; Histone H2Bq; RGD1562346; T25626 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 319190, 8349 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16778, Q64524 | |
| H2BC21 | |
| Recombinant protein corresponding to Human HIST2H2BE. Recombinant protein control fragment (Product #RP-104350). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido