Learn More
HIPK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA528787
Descripción
PA5-28787 targets HIPK1 in WB applications and shows reactivity with Human samples. The PA5-28787 immunogen is synthetic peptide corresponding to a region within amino acids 1 and 45 of Human HIPK1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
The protein encoded by this gene belongs to the Ser/Thr family of protein kinases and HIPK subfamily. It phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. Alternative splicing results in four transcript variants encoding four distinct isoforms.
Especificaciones
| HIPK1 | |
| Polyclonal | |
| Unconjugated | |
| Q86Z02 | |
| HIPK1 | |
| Synthetic peptide corresponding to a region within amino acids 1 and 45 of Human HIPK1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.38 mg/mL | |
| HIPK1 | |
| Myak, Nbak2, homeodomain interacting protein kinase 1-like protein, homeodomain-interacting protein kinase 1, nuclear body associated kinase 2b, nuclear body-associated kinase 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 204851 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.