missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NB100-654R
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
Especificaciones
| HIF-1 alpha | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated | |
| Q16665 | |
| HIF1A | |
| This HIF-1 alpha Antibody was developed against a fusion protein containing amino acids 432 - 528 of human HIF-1 alpha [Uniprot# Q16665]. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and porcine. Does not react with rat. Immunogen sequence has 93% homology to bovine and 90% homology to mouse.Mouse reactivity reported in scientific literature (PMID: 23959856) Rat reactivity reported in scientific literature (PMID: 24122166). | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
| Rabbit | |
| 93 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| 3091 | |
| Human, Mouse, Rat, Pig, Bovine | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido