missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HERV Polyclonal Antibody

Descripción
HERV Polyclonal Antibody for Western Blot, IHC (P)
HERVs are a class of multiple sclerosis associated retroviruses. Reacts with a replication competent Human Endogenous Retrovirus (HERVW) isolated from placenta. The synthetic peptide sequence used as immunogen is taken from the virus envelope polyprotein sequence. Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. Endogenous envelope proteins may have kept, lost or modified their original function during evolution. The HERVW endogenous envelope protein has retained its original fusogenic properties and participates in trophoblast fusion during placenta morphogenesis.
Especificaciones
Especificaciones
| Antígeno | HERV |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.09% sodium azide; pH 7.4 |
| génica | ERVW-1 |
| N.º de referencia del gen | Q9UQF0 |
| Alias de gen | endogenous retroviral family W, env(C7), member 1; endogenous retrovirus group W member 1; endogenous retrovirus group W, member 1; ENV; envelope glycoprotein; envelope polyprotein gPr73; envelope protein; Enverin; ENVW; Env-W; ERVW-1; ERVWE1; gp24; gp50; HERV7Q; HERV-7q; HERV-7q envelope protein; HERV-tryptophan envelope protein; HERVW; HERV-W Env glycoprotein; HERV-W envelope protein; HERV-W_7q21.2 provirus ancestral Env polyprotein; HERVWENV; HERV-W-ENV; human endogenous retrovirus W envC7-1 envelope protein; SU; Surface protein; Syncytin; syncytin A; syncytin-1; TM; Transmembrane protein |
| Símbolos de los genes | ERVW-1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?