missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: QLQGMRKGAA MLFPHTFVMA IMQPDNLGTF EIYCQAGSHR EAGMRAIYNV SQCPGHQATP RQRYQAARIY YIMAEEVEWD YCPDRSWERE WHNQSEKDSY GYIFLSNKDG LLGSRYKKAV FREYTDGTFR IPRPRTGPEE HLGILGPLI Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
B1AJX8(HEPH) is similar to an iron transport protein found in mouse. The mouse protein is similar to ceruloplasmin, a serum multi-copper ferroxidase, and is thought to be a membrane-bound protein responsible for transport of dietary iron from epithelial cells of the intestinal lumen into the circulatory system. In mouse, defects in this gene can lead to severe microcytic anemia. Three transcript variants encoding different isoforms have been described for this gene. The protein encoded by this gene is similar to an iron transport protein found in mouse. The mouse protein is similar to ceruloplasmin, a serum multi-copper ferroxidase, and is thought to be a membrane-bound protein responsible for transport of dietary iron from epithelial cells of the intestinal lumen into the circulatory system. In mouse, defects in this gene can lead to severe microcytic anemia. Two transcript variants encoding different isoforms have been described for this gene.
Especificaciones
Especificaciones
| Antígeno | HEPH |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | HEPH |
| N.º de referencia del gen | Q9BQS7 |
| Alias de gen | C130006F04Rik; Cpl; haphaestin; Heph; Hephaestin; KIAA0698; mKIAA0698; sex linked anemia; sla; UNQ2562/PRO6242 |
| Símbolos de los genes | HEPH |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?