Learn More
Hemoglobin epsilon Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Tienda Proteintech ProductosDescripción
This antibody can be used to label and isolate fetal cells from maternal blood and may be useful in prenatal diagnosis.
The epsilon globin gene (HBE) is normally expressed in the embryonic yolk sac: two epsilon chains together with two zeta chains (an alpha-like globin) constitute the embryonic hemoglobin Hb Gower I; two epsilon chains together with two alpha chains form the embryonic Hb Gower II. Both of these embryonic hemoglobins are normally supplanted by fetal, and later, adult hemoglobin. The five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5′-epsilon - G-gamma - A-gamma - delta - beta-3′
Especificaciones
Especificaciones
| Antígeno | Hemoglobin epsilon |
| Aplicaciones | Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.39 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | HBE1 |
| N.º de referencia del gen | P02100 |
| Alias de gen | Epsilon globin, HBE, HBE1, Hemoglobin epsilon chain, Hemoglobin subunit epsilon, hemoglobin, epsilon 1 |
| Símbolos de los genes | Hbe1 |
| Mostrar más |