missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hemoglobin Chimeric Recombinant Rabbit Monoclonal Antibody (2A1)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Marca:  Invitrogen™ MA548118

Código de producto. 17921001

  • 560.00€ / 200 microgramos

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Specificity: 2A1 exhibited polyreactive recognition of peptide 100-119 from the alpha subunit and peptide 100-119 frmo the beta subunit. Moreover, 2A1 bound Hb and the Hb-haptoglobin complex with equal efficiency, implying different binding sites on Hb for haptoglobin and 2A1. 2A1 was also found to distinguish between Hb and metHb. Preparation: Murine hybridomas were generated using spleen cells from aging (8-10 month old) NZB/W F1 animals. Hybridomas secreting antibodies reactive to murine hemoglobin were subcloned by limiting dilution and antibody isotypes determined using commercial reagents. Eight-week-old NZB/W F1 were immunized subcutaneously with 50 mg mHb antigen emulsified in Complete Fruends Adjuvant.

The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

Hemoglobin Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P01942
HBA, HBB
Murine hybridomas were generated using spleen cells from aging (8-10 month old) NZB/W F1 animals.
200 μg
Primary
Mouse
Antibody
IgG κ
ELISA, Western Blot, Immunocytochemistry
2A1
Unconjugated
HBB
alpha one globin; alpha-1 globin; alpha-1-globin; alpha-2 globin chain; Alpha-globin; ALPHA-GLOBIN GENE; delta globin; Haemoglobin; Hb; HBA1; HbA1c; HBA2; HBA-T3; HBH; hemoglobin alpha 1 globin chain; Hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin, alpha 1
Rabbit
Protein A
RUO
15121, 15127
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado