Learn More
HELLS Monoclonal Antibody (1D10), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA519067
Descripción
Peptide Sequence: MFGSSEKETI ELSPTGRPKR RTRKSINYSK IDDFPNELEK LISQIQPEVD RERAVVEVNI PVESEVNLKL QNIMMLLRKC CNHPYLIEYP IDPVTQEFKI DEELVTNSGK FLILDRMLPE LKKRGHKVLL FSQMTSMLDI LMDYCHLRDF NFSRLDGSMS YSEREKNMHS FNTDPEVFIF LVSTRAGGLG INLTAADTVI IYDSDWNPQS DLQAQDRCHR IGQTKPVVVY RLVTANTIDQ KIVERAAAKR KLEKLIIHKN HFKGGQSGLN LSKNFLDPKE LMELLKSRDY EREIKGSREK VISDKDLELL LDRSDLIDQM NASGPIKEKM GIFKILENSE DSSPECLF
This gene encodes a lymphoid-specific helicase. Other helicases function in processes involving DNA strand separation, including replication, repair, recombination, and transcription. This protein is thought to be involved with cellular proliferation and may play a role in leukemogenesis. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Especificaciones
| HELLS | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9NRZ9 | |
| HELLS | |
| HELLS (AAH30963, 1 a.a. ∽ 348 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1D10 | |
| Unconjugated | |
| HELLS | |
| LSH, PASG, SMARCA6, SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6, SWI/SNF2-related, matrix-associated, actin-dependent regulator of chromatin, subfamily A, member 6, lymphoid-specific helicase, proliferation-associated SNF2-like | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3070 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.