Learn More
Invitrogen™ Helicobacter Pylori OMP Recombinant Rabbit Monoclonal Antibody (RM411)
GREENER_CHOICE
Código de producto. 17866751
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
17866751 100 μL 100 microlitros
1 options
Código de producto. 17866751
Marca: Invitrogen™ MA544573
Rabbit Recombinant Monoclonal Antibody
This antibody reacts to Histone H2A.X acetylated at Lysine 9 (K9ac). No cross reactivity with non-modified Lysine 9 or other acetylated Lysines in histone H2A.
Outer membrane proteins (OMPs) ofH. pyloriplay a crucial role in the host pathogen interaction, virulence and pathogenesis. When OMPs interact with the host immune cells, OMPs stimulate the production of pro-inflammatory molecules that may result in an uncontrolled inflammation leading to the transformation of a normal cell to a cancerous cell. The Helicobacter outer membrane family of OMPs inH. pyloriconsists of four members (HomA, B, C and D).
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Helicobacter Pylori OMP
Aplicaciones ELISA
Clasificación Recombinant Monoclonal
Clon RM411
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Especie del huésped Rabbit
Inmunógeno Proteins purified from Helicobacter pylori which correspond to the full length protein.
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
Especies diana Bacteria
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG