Learn More
Invitrogen™ HECA Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA531372
Descripción
Recommended positive controls: A549, HeLa. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the homolog of the Drosophila headcase protein, a highly basic, cytoplasmic protein that regulates the re-entry of imaginal cells into the mitotic cycle during adult morphogenesis. In Drosophila, the encoded protein also inhibits terminal branching of neighboring cells during tracheal development.
Especificaciones
| HECA | |
| Polyclonal | |
| Unconjugated | |
| HECA | |
| dJ225E12.1; Gm869; H dJ225E12.1; HDC; hdc homolog, cell cycle regulator; HDCL; headcase homolog; headcase homolog (Drosophila); headcase protein homolog; HECA; hHDC; si:dkey-34f16.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51696 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.7 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UBI9 | |
| HECA | |
| Recombinant protein encompassing a sequence within the C-terminus region of human HECA. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.