missing translation for 'onlineSavingsMsg'
Learn More

Heavy chain of IgG4 Mouse anti-Human, Clone: 1A1A2, Proteintech

Código de producto. 16868255 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
Código de producto. Cantidad unitSize
16868255 150 μL 150 microlitros
16878255 20 μL 20 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16868255

Marca: Proteintech 664081IG150UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

This antibody detects the heavy chain of human IgG4. The immunogen of this antibody is the specific domain NKGLPSSIEK of IGHG4.

The regions of relatively constant sequence beyond the variable regions of immunoglobulins are termed constant regions (C regions) and are present in both the heavy and light chains. With very few exceptions, the sites of attachment for carbohydrates on immunoglobulins are located in these C regions. These regions also function to hold the variable regions together by using the disulfide bond between them. The C regions facilitate interaction with the antigen by increasing the maximum rotation of the immunoglobulin arms. Several studies have shown that a large population of patients with recurrent respiratory tract infection have low IgG4 concentrations.

Especificaciones

Antígeno Heavy chain of IgG4
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon 1A1A2
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica IGHG4
N.º de referencia del gen P01861
Alias de gen IGHG4
Símbolos de los genes IGHG4
Especie del huésped Mouse
Inmunógeno Peptide
Método de purificación Protein G
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3503
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.