missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HDEL Monoclonal Antibody (2E7), FITC
GREENER_CHOICE

Código de producto. 17837972
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17837972 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17837972 Proveedor Invitrogen™ N.º de proveedor MA545241

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

1 μg/mL of MA5-45241 was sufficient for detection of HDEL-containing proteins in 10 μg of S. cerevisiae lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 78kDa.

HSP 70 family comprises four highly conserved proteins, HSP 70, HSC 70, GRP 75 and GRP 78, which serve a variety of roles. They act as molecular chaperones, facilitating the assembly of multi-protein complexes; participate in the translocation of polypeptides across cell membranes and to the nucleus; and aid in the proper folding of nascent polypeptide chains. GRP 78 is localized in the endoplasmic reticulum (ER), where it receives imported secretory proteins and is involved in the folding and translocation of nascent peptide chains. Sorting of these proteins is dependent on a C-terminal tetrapeptide signal, usually KDEL in animal cells, and HDEL in S.cerevisiae. The 2E7 clone recognizes the C-terminal peptide HDEL, a common version of the endoplasmic reticulum retention signal found in yeast, plant, nematode and other ER proteins. 2E7 specifically stains HDEL proteins in barnyard grass, beet, cotton, mung bean, sorghum and wheat.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno HDEL
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon 2E7
Concentración 1 mg/mL
Conjugado FITC
Formulación PBS, 9.09mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
génica KdelR
N.º de referencia del gen O76767
Alias de gen BEST:CK00230; CG5183; CG5183-PA; CG5183-PB; CG5183-PC; CK00230; Dmel\CG5183; Dmel_CG5183; dmErd2; ER lumem protein retaining receptor 2; ER lumen protein retaining receptor; ER lumen protein retaining receptor 2; ER lumen protein-retaining receptor; Erd2; KDEL receptor; KdelR; KDELR1; KDEL-receptor; KdelR-PA; KdelR-PB; KdelR-PC; l(2)31Em; l(2)k00311; lethal (2) 31Em
Símbolos de los genes KdelR
Especie del huésped Mouse
Inmunógeno Raised against a synthetic HDEL peptide corresponding to the C-terminus of yeast Bip.
Método de purificación Protein G
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 34427
Especies diana Drosophila, Yeast, Plant
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.