missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDDC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Especificaciones
| Antígeno | HDDC3 |
|---|---|
| Concentración | 0.26 mg/mL |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|
16810705
|
Proteintech
21091-1-AP-20UL |
20 μL |
145.00€
20 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
16800705
|
Proteintech
21091-1-AP-150UL |
150 μL |
410.00€
150 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
HDDC3 is involved in biological processes as a hydrolyzing enzyme in starvation response.Specifications
| HDDC3 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8N4P3, Q9D114 | |
| 308758, 374659, 68695 | |
| HDDC3 Fusion Protein Ag15342 | |
| Primary | |
| -20°C | |
| HDDC3 |
| 0.26 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HD domain containing 3, HD domain containing protein 3, HDDC3 | |
| Hddc3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title