missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA596625
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Positive Samples: NIH/3T3, A-549, MCF7, Jurkat, Mouse lung, Mouse heart, Mouse brain, Rat brain; Cellular Location: Cytoplasm, Nucleus Immunogen sequence: LRQIPSAEDL ETDGGGPGQV VDDGLEHREL GHGQPEARGP APLQQHPQVL LWEQQRLAGR LPRGSTGDTV LLPLAQGGHR PLSRAQSSPA APASLSAPEP A.
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene has sequence homology to members of the histone deacetylase family. This gene is orthologous to mouse HDAC7 gene whose protein promotes repression mediated via the transcriptional corepressor SMRT. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Especificaciones
| HDAC7 | |
| Polyclonal | |
| Unconjugated | |
| HDAC7 | |
| 5830434K02Rik; HD7; HD7a; hdac4l; Hdac7; hdac-7; Hdac7a; histone deacetylase 7; Histone deacetylase 7A; mFLJ00062 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51564, 56233, 84582 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.12 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8C2B3, Q8WUI4, Q99P96 | |
| HDAC7 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human HDAC7 (XP_0115367831). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido