missing translation for 'onlineSavingsMsg'
Learn More

HDAC6 Mouse anti-Canine, Human, Rat, Clone: OTI4C5, lyophilized, TrueMAB™

Código de producto. 17681971
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
17681971 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17681971

Marca: Origene Technologies CF502030

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.

Especificaciones

Antígeno HDAC6
Aplicaciones Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon OTI4C5
Conjugado Unconjugated
Formulación PBS with 8% trehalose and no preservative; pH 7.3
génica HDAC6
N.º de referencia del gen Q9UBN7
Alias de gen CPBHM, HD6, JM21, PPP1R90
Símbolos de los genes HDAC6
Especie del huésped Mouse
Inmunógeno Full length human recombit protein of human HDAC6 produced in HEK293T cell.
Método de purificación Affinity Chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10013, 480907, 84581
Especies diana Canine, Human, Rat
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.