missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC4 Monoclonal Antibody (OTI1E1), TrueMAB™, OriGene
Description
Histone deacetylase 4 (HDAC4) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3, and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. HDAC4 is also involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D. It is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer. Mutations to the HDAC4 gene have been found in Brachydactyly-mental retardation syndrome (BDMR).
Specifications
Specifications
| Antígeno | HDAC4 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI1E1 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | HDAC4 |
| N.º de referencia del gen | P56524 |
| Alias de gen | AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?