missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCN4 Monoclonal Antibody (SHG 1E5)
GREENER_CHOICE

Código de producto. 11535792
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11535792 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11535792 Proveedor Invitrogen™ N.º de proveedor MA3903

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

MA3-903 detects HCN4 from human, mouse, and rat samples. MA3-903 has been successfully used in Western blotting and immunohistochemistry procedures. By Western blot MA3-903 detects a ~132 KDa band representing HCN4. The MA3-903 immunogen is a synthetic peptide sequence corresponding to residues S H G S L L L P P A S S P P P P Q V P Q R R G T P P L T P G R L T Q D L K L of HCN4. MA3-903 clone SHG 1E5 is a rat monoclonal hybridoma. This antibody was produced by immunizing a rat, isolating the spleen cells, creating the hybridoma and successively injecting these cells into a mouse to produce ascites.

HCN4 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. It has been shown that HCN4 channels function as receptors for sour taste, and are associated with pacemaker potential generation in the sinoatrial node.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno HCN4
Aplicaciones Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon SHG 1E5
Concentración Conc. Not Determined
Conjugado Unconjugated
Formulación Ascites with 0.05% sodium azide
génica Hcn4
N.º de referencia del gen O70507, Q9JKA7, Q9Y3Q4
Alias de gen Bcng3; BCNG-3; brain cyclic nucleotide-gated channel 3; Hcn3; Hcn4; hyperpolarization activated cyclic nucleotide gated potassium channel 4; hyperpolarization activated cyclic nucleotide-gated cation channel 4; hyperpolarization activated cyclic nucleotide-gated potassium channel 4; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4; SSS2
Símbolos de los genes Hcn4
Especie del huésped Rat
Inmunógeno Synthetic peptide sequence corresponding to residues S H G S L L L P P A S S P P P P Q V P Q R R G T P P L T P G R L T Q D L K L of HCN4.
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10021, 330953, 59266
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.