missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
HARS Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is a cytoplasmic enzyme which belongs to the class II family of aminoacyl-tRNA synthetases. The enzyme is responsible for the synthesis of histidyl-transfer RNA, which is essential for the incorporation of histidine into proteins. The gene is located in a head-to-head orientation with HARSL on chromosome five, where the homologous genes share a bidirectional promoter. The gene product is a frequent target of autoantibodies in the human autoimmune disease polymyositis/dermatomyositis.
Especificaciones
Especificaciones
| Antígeno | HARS |
| Aplicaciones | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.38 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | HARS1 |
| N.º de referencia del gen | P12081 |
| Alias de gen | CMT2W; dead end homolog 1; Dnd1; FLJ20491; HARS; HARS1; HARSL; HARSL protein; HisRS; histidine translase; histidine tRNA ligase; histidine--tRNA ligase; histidine--tRNA ligase, cytoplasmic; Histidyl-tRNA synthetase; histidyl-tRNA synthetase 1; histidyl-tRNA synthetase, cytoplasmic; HRS; im:7136719; MMHRS; Unknown (protein for IMAGE:7951312); USH3B; zgc:92215 |
| Símbolos de los genes | HARS1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
