Learn More
KLRG1 Hamster anti-Mouse, BUV395, Clone: 2F1, BD Optibuild™
Hamster Monoclonal Antibody
Marca: BD Biosciences 740279
Descripción
The 2F1 monoclonal antibody specifically binds to KLRG1 ( K iller cell L ectin-like R eceptor mmunoreceptor T yrosine-based I nhibitory M otif); its ligand has not been identified KLRG1 is expressed mainly as a homodimeric molecule consisting of two N-glycosylated subunits of approximately 30-38 kDa. The level of KLRG1 expression is reduced in MHC class I-deficient mice, although direct binding of KLRG1 to MHC class I antigens could not be detected. Crosslinking of KLRG1 by 2F1 mAb reduces TCR-mediated Ca++ mobiliza tion and cytotoxic responses (but not IFN-γproduction) by CD8+ T cells and inhibits IFN-γand TNF production and redirected lysis by NK cells.
Especificaciones
| KLRG1 | |
| Monoclonal | |
| 0.2mg/mL | |
| Klrg1; Killer cell lectin-like receptor subfamily G member 1; MAFA; 2F1 Ag | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2 κ |
| Flow Cytometry | |
| 2F1 | |
| Brilliant Ultraviolet 395 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| A-LAK from C57BL/6 mice | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only