Descripción
- Tear-, flame-, lint- and abrasion-resistant
- POWERGUARD technology for enhanced microbial barrier protection
- Pre-vacuum steam sterilant penetration
- Ethylene Oxide (EO) sterilant penetration and residuals
- Maintenance of package integrity (MPI) to meet FDA requirements
- Instructions for Use that meet FDA guidelines
- Class 1 flammability rating
- Low linting
- Made in the USA
Especificaciones
Especificaciones
| Esterilizables en autoclave | Autoclavable |
| Certificaciones/conformidad | Meets NFPA Class 1 |
| Color | Azul |
| Para utilizar con (equipo) | Basin sets, instrument sets, procedure trays and large linen packs (6 to 9 lb.) |
| Material | SMS Fabric (H300) |
| Esterilidad | No estéril |
| Sin látex | Sí |
| Dimensiones (L x An) | 76 x 76 cm (30 x 30 in) |
| Longitud (imperial) | 30 in |
| Anchura (imperial) | 30 in |
| Mostrar más |