Learn More
Gilac™ HACCP Cook Chill Crate
Descripción
- 600 x 400 perforated crate.
- Ideal for commercial kitchen use and the cold chain process.
- Provides ventilation.
- Stacking and nesting when half turned.
- Large perforations, meaning that temperatures can be brought down quickly and it is easy to clean.
- Crates available in 5 HACCP colors.
- Tracking management thanks to a contents identification label (IML): In mold labelling process whereby the label is injection-molded into the plastic rendering it permanent and rewritable.
- Use with a water erasable marking pen.
Especificaciones
Especificaciones
| Capacidad (imperial) | 9.8 gal. |
| Capacidad (métrico) | 37 L |
| Tipo de tapón | None |
| Color | Amarillo |
| Material | HDPE |
| Forma | Rectangulares |
| Tipo de producto | Crate |
| Dimensiones (Largo x Ancho x Alto) | 600 x 400 x 197 mm |
| Longitud (métrico) | 600 mm |
| Longitud (imperial) | 23.6 in. |
| Mostrar más |