missing translation for 'onlineSavingsMsg'
Learn More

HAAO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Código de producto. 16850694 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
Código de producto. Cantidad unitSize
16850694 20 μL 20 microlitros
16840694 150 μL 150 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16850694

Marca: Proteintech 127911AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

HAAO (3-Hydroxyanthranilate 3, 4-dioxygenase) is a monomeric cytosolic protein of the family of intramolecular dioxygenases containing non-heme ferrous iron. It is widely distributed in peripheral organs, such as liver and kidney, and is present in low amounts in the central nervous system. This enzyme participates in tryptophan metabolism. It employs one cofactor, iron. HAAO catalyzes the synthesis of quinolinic acid (QUIN) from 3-hydroxyanthranilic acid. QUIN is an excitotoxin whose toxicity is mediated by its ability to activate glutamate N-methyl-D-aspartate receptors. Increased cerebral levels of QUIN may participate in the pathogenesis of neurological and inflammatory disorders. HAAO has been suggested to play a role in disorders associated with altered tissue levels of QUIN. Furthermore, recent study shows that HAAO are excellent candidate biomarkers for detecting ovarian cancer.

Especificaciones

Antígeno HAAO
Aplicaciones Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.29 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica HAAO
N.º de referencia del gen P46952, P46953, Q78JT3
Alias de gen 3 HAO, HAAO, HAD, HAO
Símbolos de los genes Haao
Especie del huésped Rabbit
Inmunógeno HAAO Fusion Protein Ag3511
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 107766, 23498, 56823
Especies diana Mouse, Human, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.