missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ H4K20me1 Monoclonal Antibody (5E10-D8), ChIP-Verified
GREENER_CHOICE

Código de producto. 15314341
Change view
Click to view available options
Cantidad:
100 μL
20 μg
Tamaño de la unidad:
100 microlitros
20 microgramos
Código de producto. Cantidad unitSize
15314341 100 μL 100 microlitros
17145113 20 μg 20 microgramos
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15314341

Marca: Invitrogen™ MA518067

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

This antibody is specific for K20 mono-methylated histone H4 and does not cross-react with histone 3 or tri-, di-, or unmethylated histone H4. This antibody is predicted to react with Xenopus laevis, Arabidopsis thaliana, Fruit fly (Drosophila melanogast), and most mammals based on sequence homology. This target has a predicted molecular weight of 11 kDa. Actual Western blot results show a band at 13 kDa. This can be blocked using mono-methyl K20 Histone H4 peptide. Click here for Master Lot linking information.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno H4K20me1
Aplicaciones ChIP Assay, Dot Blot, ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon 5E10-D8
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.1% sodium azide
génica H4C1
N.º de referencia del gen P62805
Alias de gen BcDNA:RH52884; CELE_T10C6.11; CG31611; CG3379; CG3379-PA; CG3379-PB; CG3379-PC; CG3379-PD; CG33869; CG33871; CG33873; CG33875; CG33877; CG33879; CG33881; CG33883; CG33885; CG33887; CG33889; CG33891; CG33893; CG33895; CG33897; CG33899; CG33901; CG33903; CG33905; CG33907; CG33909; Dmel\CG3379; Dmel_CG3379; F07B7.11; F07B7.4; F45F2.12; FBtr0082962; FO108; H4; H4 clustered histone 11; H4 clustered histone 2; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K20; H4.1; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/N; H4/O; H4/p; H4-12; H4-16; H4-53; H4C1; H4C11; H4C12; H4C13; H4c14; H4C15; H4C2; H4c3; H4c4; H4C5; H4c6; H4c8; H4C9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K20; H4K20me; H4Lys20me; H4Lys20me1; H4M; H4r; his-20; his-22; His4; his-4; His4:CG31611; His4:CG33869; His4:CG33871; His4:CG33873; His4:CG33875; His4:CG33877; His4:CG33879; His4:CG33881; His4:CG33883; His4:CG33885; His4:CG33887; His4:CG33889; His4:CG33891; His4:CG33893; His4:CG33895; His4:CG33897; His4:CG33899; His4:CG33901; His4:CG33903; His4:CG33905; His4:CG33907; His4:CG33909; His4-88CD; His4r; His4r-PA; His4r-PB; His4r-PC; His4r-PD; his-52; his-54; his-8; HIST1H4; hist1h4a; hist1h4a.L; Hist1h4b; Hist1h4c; Hist1h4d; HIST1H4E; Hist1h4f; HIST1H4H; HIST1H4I; HIST1H4J; Hist1h4k; HIST1H4L; Hist1h4m; Hist2h4; HIST2H4A; HIST2H4B; HIST4H4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 1, H4l; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4a L homeolog; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; Histone H2B 2; histone H3; histone H4; Histone H4 replacement; histone H4.1; Histone H4A; Histone H4K20me1; histone IV, family 2; K06C4.12; K06C4.4; LOC102161782; methyl Histone 3; methyl Histone H3; Methyl-H4 K20; Methyl-H4 Lys20; Methyl-Histone H4 K20; Methyl-Histone H4 Lys20; Mono-methyl H4; Mono-methyl Histone H4; RH52884p; T10C6.11; X04652; XELAEV_18001418mg
Símbolos de los genes H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9
Especie del huésped Mouse
Inmunógeno Synthetic peptide within Human Histone H4 (mono methyl K20)(Immunogen sequence: 15-AKRHR(Kme)VLRDN-25 (branched)).
Método de purificación Protein G
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 121504, 8294, 8359, 8363, 8366, 8367, 8370
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.