missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-2Kk Mouse anti-Mouse, BUV395, Clone: AF3-12.1, BD Optibuild™
Mouse Monoclonal Antibody
Marca: BD Biosciences 743940
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The AF3-12.1 antibody specifically recognizes the H-2K[k] MHC class I alloantigen. Reactivity with other haplotypes( e.g., b, d, f, j, p, q, r, s, u, v /) has not been observed.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Especificaciones
| H-2Kk | |
| AF3-12.1 | |
| Brilliant Ultraviolet 395 | |
| H2-K, H-2 class I histocompatibility antigen K, H-2K.23 | |
| A/J mouse splenocytes | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido