missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GZF1 Polyclonal Antibody
Descripción
A suggested positive control is human heart tissue lysate. PA5-34375 can be used with blocking peptide PEP-1418.
The GDNF-inducible zinc finger protein 1 (GZF1) is a sequence-specific transcriptional repressor with a BTB/POZ domain and ten zinc finger motifs whose expression is required for renal branching morphogenesis during kidney development. GZF1 binds to the 5'regulatory region of the homeodomain protein HOXA10, suggesting that GZF1 may play a role in morphogenesis other than kidney development. Recent experiments have indicated that GZF1 associates with nucleolin and this association is mediated by the first four zinc finger motifs of GZF1. It is thought that Nucleolin modulates the subcellular localization of GZF1 as well as its transcriptional repressor activity.
Especificaciones
Especificaciones
| Antígeno | GZF1 |
| Aplicaciones | Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.02% sodium azide |
| génica | GZF1 |
| N.º de referencia del gen | Q9H116 |
| Alias de gen | GDNF inducible zinc finger protein 1; GDNF-inducible zinc finger protein 1; GZF1; ZBTB23; Zinc finger and BTB domain-containing protein 23; zinc finger protein 336; ZNF336 |
| Símbolos de los genes | GZF1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?