missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2IRD2 Polyclonal Antibody

Descripción
GTF2IRD2 Polyclonal Antibody for Western Blot, Flow
GTD2A is unusual in that its coding sequence is mostly derived from Charlie8 repeat elements. However, there is mRNA and EST evidence to suggest that this gene is transcribed, and the encoded protein has a homolog in mouse, with which it shares 78% sequence identity. The exact function of this gene product is not known. It is inferred to be a transcription factor based on the presence of GTF2I-like repeats (containing helix-loop-helix motifs), also found in other proteins such as GTF2IRD1 and GTF2I. These three genes are clustered together on chr 7q11.23.
Especificaciones
Especificaciones
| Antígeno | GTF2IRD2 |
| Aplicaciones | Flow Cytometry, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.09% sodium azide; pH 7.4 |
| génica | GTF2IRD2 |
| N.º de referencia del gen | Q86UP8 |
| Alias de gen | FP630; general transcription factor II i repeat domain 2 alpha; general transcription factor II-I repeat domain-containing protein 2A; GTF2I repeat domain containing 2; GTF2I repeat domain-containing protein 2, alpha; GTF2I repeat domain-containing protein 2A; GTF2IRD2; GTF2IRD2 alpha; GTF2IRD2A; Transcription factor GTF2IRD2-alpha |
| Símbolos de los genes | GTF2IRD2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?