missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2IRD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17052-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Williams-Beuren syndrome (WBS) is a developmental disorder caused by the hemizygous microdeletion on chromosome 7q11.23. WBS is an autosomal domit genetic condition that is characterized by physical, cognitive and behavioral traits. The physical traits associated with WBS include facial dysmorphology, vascular stenoses, growth deficiencies, dental anomalies and neurologic and musculoskeletal abnormalities. Mild retardation, a weakness in visual-spatial skills, anxiety and a short attention span are typical cognitive and behavioral traits of WBS patients. The WBSCR11 gene is located within the WBS deletion and may contribute to the developmental symptoms found in WBS because of a loss of the encoded transcription factor. WBSCR11 is also designated GRF2IRD1, GTF3, Cream1 and MusTRD1 in human and BEN in mouse, due to slight differences in gene structure. WBSCR11 is expressed in all adult tissues as several variants and has discrete spatial and temporal expression during embryogenesis.Especificaciones
| GTF2IRD1 | |
| Polyclonal | |
| Unconjugated | |
| GTF2IRD1 | |
| BEN, CREAM1, GTF2IRD1, GTF3, hMusTRD1alpha1, MUSTRD1, MusTRD1/BEN, RBAP2, USE B1 binding protein, WBS, WBSCR11, WBSCR12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 246770, 57080, 9569 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UHL9 | |
| Gtf2ird1 | |
| GTF2IRD1 Fusion Protein Ag10718 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido