missing translation for 'onlineSavingsMsg'
Learn More

GSTT2 Monoclonal Antibody (OTI7A12), TrueMAB™, OriGene

Product Code. 17565361
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Unit Size:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
17565361 100 μL 100 microlitros
17585361 30 μL 30 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17565361 Supplier Origene Technologies Supplier No. TA501858

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. Has a sulfatase activity.

Specifications

Antígeno GSTT2
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon OTI7A12
Concentración 0.48 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica GSTT2
N.º de referencia del gen P0CG29
Alias de gen GSTT2B
Símbolos de los genes GSTT2
Especie del huésped Mouse
Inmunógeno Full length human recombit protein of human GSTT2 produced in HEK293T cell.
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2953
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.