Learn More
STEMCELL Technologies GSK621
Descripción
- GSK621 is a specific agonist of AMP-activated protein kinase (AMPK; Sujobert et al.)
- Increases AMPKα T172 phosphorylation, a marker of AMPK activation, in acute myeloid leukemia (AML) cells and primary AML samples (Sujobert et al.)
- Displays selective cytotoxicity (IC50 = 13 - 30 μM) by activating autophagy via the eIF2ɑ signaling pathway independent of mTORC1 activation in AML cell lines but not in normal hematopoietic progenitor cells (Sujobert et al.)
- Inhibits human melanoma cell survival and proliferation (Chen et al.)
Especificaciones
Especificaciones
| CAS | 1346607-05-3 |
| Cantidad | 1 mg |
| Fórmula molecular | C26H20ClN3O5 |
| Pureza | ≥98% |