missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSK3 alpha Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA585988
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GSK3 alpha Polyclonal Antibody for ICC/IF, Western Blot
Glycogen synthase kinase 3 alpha (GSK3A) is a multifunctional protein serine/threonine kinase implicated in the control of several regulatory proteins and transcription factors. (GSK3A Alias: DKFZp686D0638). Human GSTs can be divided into five main classes: alpha, mu, pi, theta, and zeta. The theta class includes GSTT1 and GSTT2. The GSTT1 and GSTT2 share 55% amino acid sequence identity and both of them were claimed to have an important role in human carcinogenesis. The GSTT1 gene is located approximately 50kb away from the GSTT2 gene. The GSTT1 and GSTT2 genes have a similar structure, being composed of five exons with identical exon/intron boundaries.
Especificaciones
| GSK3 alpha | |
| Polyclonal | |
| Unconjugated | |
| Gsk3a | |
| 2700086H06Rik; DKFZp686D0638; FA; factor A; glycogen synthase kinase 3 alpha; glycogen synthase kinase 3 alpha b; glycogen synthase kinase 3 alpha b; glycogen synthase kinase-3 alpha; glycogen synthase kinase-3 alpha; GSK3; GSK-3 alpha; GSK-3[a]; GSK-3[b]; gsk3a; gsk3ab; gsk-3-alpha; OTTHUMP00000198368; Serine/threonine-protein kinase GSK3A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2931 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P49840 | |
| Gsk3a | |
| Synthetic peptide within Human GSK3 alpha aa 434-483. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?