missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-3 beta, p Ser9 Antibody (609739), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB25062W
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GSK-3 beta Monoclonal antibody specifically detects GSK-3 beta in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Especificaciones
| GSK-3 beta | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 609739 | |
| Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform | |
| Phosphopeptide containing the human GSK-3 beta S9 site | |
| 0.1 mL | |
| Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 2932 | |
| Store at 4°C in the dark. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido