missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
This antibody was generated against the C-terminal region of full-length HSP90b1.
GRP94 is a 803 amino acid protein belonging to the heat shock protein 90 family. It acts as a molecular chaperone that functions in the processing and transport of secreted proteins. GRP94 and its N-terminal fragment stimulates CTL expansion and maturation of human monocyte-derived dendritic cells (MDDC). It plays a central role in innate as well as acquired immunity, maturation and chemotaxis of dendritic cells, Ab production, cross-priming, as a potential marker in breast cancer and is a peptide acceptor in endoplasmic reticulum and an accessory to peptide loading of MHC class I molecules. Expression of GRP94 suppressed A23187-induced apoptosis and stabilized calcium homeostasis. GRP94 is expressed in melanoma or liver metastases of colon carcinoma cells, human gastric carcinoma BGC-823 cells.
Especificaciones
Especificaciones
| Antígeno | GRP94 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry, Immunocytochemistry, Immunoprecipitation |
| Clasificación | Polyclonal |
| Concentración | 0.24 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | HSP90B1 |
| N.º de referencia del gen | P08113, P14625, Q66HD0 |
| Alias de gen | ECGP, Endoplasmin, GP96, gp96 homolog, GRP 94, GRP94, HSP90B1, TRA1, Tumor rejection antigen 1 |
| Símbolos de los genes | Hsp90b1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?