Learn More
TRIM55, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TRIM55.
Marca: Abnova PAB6355.100ug
Descripción
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein associates transiently with microtubules, myosin, and titin during muscle sarcomere assembly. It may act as a transient adaptor and plays a regulatory role in the assembly of sarcomeres. Four alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq
Sequence: C-HIFSFSWLNSLNEEspecificaciones
| TRIM55 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
| TRIM55 | |
| TRIM55 | |
| A synthetic peptide corresponding to human TRIM55. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize isoforms 1, 3 and 4 (NP_908973.1, NP_908974.1and NP_908975.1 resp.). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of TRIM55. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| MURF-2/RNF29 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 84675 | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only