Learn More
TPH2 Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TPH2.
Marca: Abnova PAB6927.100ug
Descripción
This gene encodes a member of the pterin-dependent aromatic acid hydroxylase family. The encoded protein catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. The human genome contains two related tryptophan hydroxylases, one on chromosome 11p15-p14 and one on chromosome 12q21. This gene is expressed predominantly in the brain stem. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression. [provided by RefSeq
Sequence: C-NKPNSGKNDDKGNKEspecificaciones
| TPH2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
| TPH2 | |
| TPH2 | |
| A synthetic peptide corresponding to human TPH2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of TPH2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| FLJ37295/MGC138871/MGC138872/NTPH | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 121278 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only