missing translation for 'onlineSavingsMsg'
Learn More

TBP (isoform 1), Goat, Polyclonal Antibody, Abnova™

Código de producto. 16034887
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
16034887 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16034887

Marca: Abnova PAB24603.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat polyclonal antibody raised against synthetic peptide of TBP (isoform 1).

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes TBP, the TATA-binding protein. A distinctive feature of TBP is a long string of glutamines in the N-terminal. This region of the protein modulates the DNA binding activity of the C terminus, and modulation of DNA binding affects the rate of transcription complex formation and initiation of transcription. Mutations that expand the number of CAG repeats encoding this polyglutamine tract, and thus increase the length of the polyglutamine string, are associated with spinocerebellar ataxia 17, a neurodegenerative disorder classified as a polyglutamine disease. [provided by RefSeq

Sequence: DQNNSLPPYAQC

Especificaciones

Antígeno TBP (isoform1)
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Descripción Goat polyclonal antibody raised against synthetic peptide of TBP (isoform1).
Dilución ELISA (1:64000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulación In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
génica TBP
Alias de gen GTF2D/GTF2D1/MGC117320/MGC126054/MGC126055/SCA17/TFIID
Símbolos de los genes TBP
Especie del huésped Goat
Inmunógeno A synthetic peptide corresponding to N-terminus of human TBP (isoform1).
Método de purificación Antigen affinity purification
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6908
Especificidad de la prueba This antibody is expected to recognize reported isoform 1(NP_003185.1) only.
Especies diana Human
Contenido y almacenamiento Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.