missing translation for 'onlineSavingsMsg'
Learn More

SNF4 (Candida), Goat, Polyclonal Antibody, Abnova™

Código de producto. 16243840
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
16243840 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 16243840

Brand: Abnova PAB28042.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Goat polyclonal antibody raised against native SNF4 (Candida).

Specifications

Antígeno SNF4 (Candida)
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Descripción Goat polyclonal antibody raised against native SNF4 (Candida).
Dilución ELISA (1:5000 - 1:25000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user.
Formulación Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
génica SNF4
Símbolos de los genes SNF4
Especie del huésped Goat
Inmunógeno Native purified Candida SNF4.
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3640338
Especies diana Bacteria
Contenido y almacenamiento Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Formulario Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.