Learn More
SERPINI2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SERPINI2.
Marca: Abnova PAB6764.100ug
Descripción
The protein encoded by this gene is a member of the serine protease inhibitor (serpin) superfamily made up of proteins which play central roles in the regulation of a wide variety of physiological processes, including coagulation, fibrinolysis, development, malignancy and inflammation. The gene product may have a role in a growth-control, possibly growth-suppressing pathway and, when impaired, may be involved in pancreatic carcinogenesis. The protein is a member of the plasminogen activator inhibitor-1 family, a subset of the serpin superfamily whose members act as tissue-specific tPA inhibitors. Two alternatively spliced transcript variants encoding distinct protein isoforms have been found for this gene. [provided by RefSeq
Sequence: C-KGDWKQKFRKEDTQEspecificaciones
| SERPINI2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.3-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3 ug/mL) The optimal working dilution should be determined by the end user. | |
| SERPINI2 | |
| SERPINI2 | |
| A synthetic peptide corresponding to internal region of human SERPINI2. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_006208.1, NP_00101230.3.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of SERPINI2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| MEPI/PANCPIN/PI14/TSA2004 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5276 | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only