missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTN, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PTN.
Marca: Abnova PAB19706.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Sequence: CGKLTKPKPQAESKKEspecificaciones
| PTN | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of PTN. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| HARP/HBGF8/HBNF/NEGF1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5764 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| PTN | |
| PTN | |
| A synthetic peptide corresponding to C-terminus of human PTN. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction