Learn More
PTGER4, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PTGER4.
Marca: Abnova PAB6969.100ug
Descripción
The protein encoded by this gene is a member of the G-protein coupled receptor family. This protein is one of four receptors identified for prostaglandin E2 (PGE2). This receptor can activate T-cell factor signaling. It has been shown to mediate PGE2 induced expression of early growth response 1 (EGR1), regulate the level and stability of cyclooxygenase-2 mRNA, and lead to the phosphorylation of glycogen synthase kinase-3. Knockout studies in mice suggest that this receptor may be involved in the neonatal adaptation of circulatory system, osteoporosis, as well as initiation of skin immune responses. [provided by RefSeq
Sequence: C-LEREVSKNPDLQAEspecificaciones
| PTGER4 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| PTGER4 | |
| PTGER4 | |
| A synthetic peptide corresponding to human PTGER4. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PTGER4. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| EP4/EP4R/MGC126583 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5734 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only