Learn More
Invitrogen™ Goat anti-Mouse IgG1 Secondary Antibody, PE-Texas Red
Código de producto. 10291853
Change view
Click to view available options
Cantidad:
500 μL
Tamaño de la unidad:
cada uno
10291853 500 μL cada uno
1 options
Código de producto. 10291853
Marca: Invitrogen™ M32017
Goat Polyclonal Secondary Antibody
Goat anti-Mouse IgG1 Secondary Antibody for Flow
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Mouse IgG1
Aplicaciones Flow Cytometry
Clasificación Polyclonal
Conjugado PE-Texas Red
Formulación PBS with 0.1% sodium azide
Especie del huésped Goat
Inmunógeno IgG gamma 1.
Método de purificación Purified
Cantidad 500 μL
Estado normativo RUO
Primario o secundario Secondary
Especies diana Mouse
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG